Merck
  • Inhibition of esterase activity and an undercounting of circulating monocytes in a population of production workers.
Inhibition of esterase activity and an undercounting of circulating monocytes in a population of production workers.
Journal of occupational medicine. : official publication of the Industrial Medical Association (1986-03-01)
M S Levine, N L Fox, B Thompson, W Taylor, A C Darlington, J Van der Hoeden, E A Emmett, W Rutten
ABSTRACT
A cross-sectional review of routine employee health status examinations has identified an undercounting of circulating monocytes associated with an inhibition of a surface monocyte esterase, alpha-naphthyl butyrate esterase. It is postulated that this inhibition is associated with an organophosphate used in the production process. Correlation with routine measures of health status are presented.
Sigma-Aldrich
1-Naphthyl butyrate, esterase substrate