Merck
[Peculiarities of thyroid pathology in the childhood].
Likars'ka sprava (2013-03-29)
V N Buriak, E S Murashko
ABSTRACT
The paper deals with etiopathogenetic and clinical peculiarities, and also therapeutic methods during the pathological processes in the thyroid system in the childhood. The most common of these processes is the diffuse nontoxic goiter, which results in abnormality of metabolic processes regulation in the child's growing organism and often signals to the beginning of many thyropathies.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Supelco
3,3′,5-Triiodo-L-thyronine (T3) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®