Merck
  • Rapid one-pot propargylamine synthesis by plasmon mediated catalysis with gold nanoparticles on ZnO under ambient conditions.
Rapid one-pot propargylamine synthesis by plasmon mediated catalysis with gold nanoparticles on ZnO under ambient conditions.
Chemical communications (Cambridge, England) (2013-01-24)
María González-Béjar, Kate Peters, Geniece L Hallett-Tapley, Michel Grenier, Juan C Scaiano
ABSTRACT
Surface plasmon excitation of gold nanoparticles on ZnO in the presence of an aldehyde, an amine and phenylacetylene led to rapid and selective formation of propargylamines with good yields (50-95%) at room temperature. Plasmon mediated catalysis is the best available route for this ternary coupling.
Sigma-Aldrich
Propargylamine, 98%
Sigma-Aldrich
Propargylamine hydrochloride, 95%
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Pargyline hydrochloride