Merck
  • Altering the rest interval during high-intensity interval training does not affect muscle or performance adaptations.
Altering the rest interval during high-intensity interval training does not affect muscle or performance adaptations.
Experimental physiology (2012-08-28)
Johann Edge, Nir Eynon, Michael J McKenna, Craig A Goodman, Roger C Harris, David J Bishop
ABSTRACT
It has been hypothesized that exercise-induced changes in metabolites and ions are crucial in the adaptation of contracting muscle. We tested this hypothesis by comparing adaptations to two different interval-training protocols (differing only in the rest duration between intervals), which provoked different perturbations in muscle metabolites and acid-base status. Prior to and immediately after training, 12 women performed the following tests: (1) a graded exercise test to determine peak oxygen uptake (V(O2)); (2) a high-intensity exercise bout (followed 60 s later by a repeated-sprint-ability test; and (3) a repeat of the high-intensity exercise bout alone with muscle biopsies pre-exercise, immediately postexercise and after 60 s of recovery. Subjects performed 5 weeks (3 days per week) of training, with either a short (1 min; HIT-1) or a long rest period (3 min; HIT-3) between intervals; training intensity and volume were matched. Muscle [H(+)] (155 ± 15 versus 125 ± 8 nmol l(-1); P < 0.05) and muscle lactate content (84.2 ± 7.9 versus 46.9 ± 3.1 mmol (g wet weight)(-1)) were both higher after HIT-1, while muscle phosphocreatine (PCr) content (52.8 ± 8.3 versus 63.4 ± 9.8 mmol (g wet weight)(-1)) was lower. There were no significant differences between the two groups regarding the increases in , repeated-sprint performance or muscle Na(+),K(+)-ATPase content. Following training, both groups had a significant decrease in postexercise muscle [H(+)] and lactate content, but not postexercise ATP or PCr. Postexercise PCr resynthesis increased following both training methods. In conclusion, intense interval training results in marked improvements in muscle Na(+),K(+)-ATPase content, PCr resynthesis and . However, manipulation of the rest period during intense interval training did not affect these changes.
Sigma-Aldrich
L-Carnosine, ~99%, crystalline