Merck
  • Estrogens and synthetic androgens in manure slurry from trenbolone acetate/estradiol implanted cattle and in waste-receiving lagoons used for irrigation.
Estrogens and synthetic androgens in manure slurry from trenbolone acetate/estradiol implanted cattle and in waste-receiving lagoons used for irrigation.
Chemosphere (2012-07-17)
Bushra Khan, Linda S Lee
ABSTRACT
The increasing size of concentrated animal feeding operations has led to a concomitant increase in the land-application of manure, which has spawned research on the concentrations and environmental risk assessment of natural and synthetic hormones in animal manures. 17β-Trenbolone acetate (TBA) is widely used in the United States for improving daily gains in beef cattle and is often administered in combination with 17β-estradiol (17β-E2). Trenbolone (TB) and E2 isomers and their metabolites were quantified in manure collection pits and lagoon effluent from beef cattle implanted with the commercial anabolic preparation Ravoler-S (containing 140 mg 17β-trenbolone acetate and 28 mg 17β-E2). Manure pit and lagoon effluent samples were collected weekly for 9 weeks post implanting and analyzed using reverse-phase liquid chromatography tandem mass spectrometry. 17α-TB was the most abundant androgen with the highest concentration observed 2 weeks post implant. 17β-TB and trendione peaked at the end of week 2 and 4, respectively. For the estrogens, the highest concentrations for estrone (E1), estriol (E3), and 17α-E2 were observed after week 4, 6, and 8, respectively. 17β-E2 concentrations were the lowest of the estrogens and erratic over time. In lagoon water, which is used for irrigation, 17α-TB and E1 had the highest detected hormone concentrations (1.53 and 1.72 μg L(-1), respectively). Assuming a 1-2 order dilution during transport to surface water, these hormone levels could lead to concentrations in receiving waters that exceed some of the lowest observable effect levels (LOELs) reported for hormones (e.g., 0.01-0.03 μg L(-1)).
Product Description
Supelco
Trenbolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Trenbolone, analytical standard, for drug analysis