Merck
  • Bradykinin receptor antagonists containing N-substituted amino acids: in vitro and in vivo B(2) and B(1) receptor antagonist activity.
Bradykinin receptor antagonists containing N-substituted amino acids: in vitro and in vivo B(2) and B(1) receptor antagonist activity.
Journal of medicinal chemistry (1996-03-29)
V S Goodfellow, M V Marathe, K G Kuhlman, T D Fitzpatrick, D Cuadrado, W Hanson, J S Zuzack, S E Ross, M Wieczorek, M Burkard, E T Whalley
ABSTRACT
We report a systematic probing of the structural requirements of the bradykinin (BK) type 2 (B(2)) receptor for antagonist activity by incorporating N-alkyl-amino acid residues at positions 7 and 8 of a potent antagonist sequence. Compound 1 (D-Arg(0)-Arg(1)-Pro(2)-Hyp(3)-Gly(4)-Thi(5)-Ser(6)-D-Tic(7)-N-Chg (8)-Arg(9), CP-0597)(1,2) is a potent (pA(2) = 9.3, rat uterus; pK(i) = 9.62, binding, human receptor clone) B(2) receptor antagonist devoid of in vitro B(1) antagonist activity (rabbit aorta). Compound 1 exhibits high potency (ED(50) = 29.2 pmol/kg/min, iv, rabbit) and duration of action when tested in models for in vivo B(2) antagonist activity. Although devoid of activity in a classic B(1) isolated tissue assay, B(1) antagonist activity for 1 was demonstrated in vivo, in a LPS-treated, inducible BK(1) receptor rabbit blood pressure model (ED(50) = 1.7 nmol/kg/min). D-Arg(0) of 1 can be formally replaced by an achiral arginine surrogate, without significant loss in antagonist potency on rat uterus (compound 11, B(2) pA(2) = 9.1). Antagonist 13 (Hyp(2), Nchg(8)), pK(i) = 10.2, and agonist 4 (N-methylcyclohexyl-Gly(8)), pK(i) = 10.1, also exhibited substantial binding to guinea pig ileum membrane receptors as well as a human B(2) receptor clone. Very minor structural changes in the N-alkyl amino acid residues in positions 7 and 8 can modify the activity of this class of compounds from being extremely potent antagonists to tight binding partial or full agonists. These studies have resulted in a series of compounds containing inexpensive amino acid residues but which produce broad spectrum BK receptor blocking potency and exceptional in vivo duration of action.