Merck
  • The amphetamine-chlordiazepoxide mixture, a pharmacological screen for mood stabilizers, does not enhance amphetamine-induced disruption of prepulse inhibition.
The amphetamine-chlordiazepoxide mixture, a pharmacological screen for mood stabilizers, does not enhance amphetamine-induced disruption of prepulse inhibition.
Behavioural brain research (2011-08-09)
Tessa N Douma, Adam Kolarz, Ynte Postma, Berend Olivier, Lucianne Groenink
ABSTRACT
In rodents, administration of a mixture of the psychostimulant d-amphetamine and the benzodiazepine chlordiazepoxide results in supra-additive hyperlocomotion, a phenomenon used to identify mood stabilizers. In an attempt to determine whether the d-amphetamine/chlordiazepoxide assay could extend to other behaviors that are affected in mania, we evaluated the effects of the mixture on prepulse inhibition. In addition, we combined chlordiazepoxide with the selective dopamine reuptake inhibitor GBR 12909 or the noradrenergic stimulant (-) ephedrine, and tested these alternative mixtures in locomotor activity and prepulse inhibition tests. Chlordiazepoxide (3mg/kg) robustly potentiated amphetamine-induced hyperactivity, but did not change the amphetamine-induced disruption of prepulse inhibition. This indicates that the d-amphetamine-chlordiazepoxide-induced hyperlocomotion does not extend to other dopamine-driven behaviors. GBR 12909 (16mg/kg) and (-) ephedrine (50mg/kg) both enhanced locomotor activity and disrupted PPI, but combined treatment of either of these compounds with chlordiazepoxide had no significant additive effect on locomotor activity or prepulse inhibition. These findings suggest that the effect of the d-amphetamine/chlordiazepoxide mixture cannot be accounted for by the dopamine enhancing properties of amphetamine alone. Last, valproic acid (120-240mg/kg) did not reduce the GBR-induced hyperactivity. Therefore, further pharmacological evaluation of GBR 12909-induced hyperactivity is warranted to determine its pharmacological potential to model mania-like behavior. Based on the current results, it is concluded that the utility of the pharmacological d-amphetamine/chlordiazepoxide assay as a tool to study brain mechanisms relevant to mania is limited.
Sigma-Aldrich
GBR 12909 dihydrochloride, solid, ≥98% (HPLC)
Supelco
Chlordiazepoxide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®