Merck
Solid thoron source preparation in a porous mineral matrix.
Radiation protection dosimetry (2010-09-11)
V Jobbágy, B Bety-Denissa
ABSTRACT
Thoron and progeny are decay products of (232)Th with a great impact on human health. The release of thoron gas from the mining and milling of thorite, monazite and other major thorium ores has been recognised as a potential radiological health hazard. For precise measurements, calibration is a very important factor. This paper describes a cheap and easy way of producing a stable thoron source made of thorium nitrate packed in a porous clay mineral matrix used as (220)Rn generator. The source should have a small spherical shape and be fired at 600°C; this will lead to a great pore volume, necessary for the thoron gas. High importance should be given to the water uptake. The exhalation power of (220)Rn was measured using a Lucas scintillation cell. Experimental efficiency values obtained ranged between 0.16 and 1.44 %.