Merck
  • Formation of organic acids from the gas-phase ozonolysis of terpinolene.
Formation of organic acids from the gas-phase ozonolysis of terpinolene.
Physical chemistry chemical physics : PCCP (2009-05-22)
Yan Ma, George Marston
ABSTRACT
Gas-phase ozonolysis of terpinolene was studied in static chamber experiments using gas chromatography coupled to mass spectrometric and flame ionisation detection to separate and detect products. Two isomers of C(7)-diacids and three isomers of C(7)-aldehydic acids were identified in the condensed phase after derivatisation. Possible mechanisms of formation of these acids were investigated using different OH radical scavengers and relative humidities, and were compared to those reported earlier for the ozonolysis of beta-pinene. In addition, branching ratios for some of the individual reaction steps, e.g. the branching ratio between the two hydroperoxide channels of the C(7)-CI, were deduced from the quantitative product yield data. Branching ratios for POZ decomposition and the stabilisation/decomposition of the C(7-)CI were also obtained from measurements of the C(7) primary carbonyl product.
Sigma-Aldrich
Terpinolene, ≥95%, FG
Sigma-Aldrich
Terpinolene, technical, ≥85% (GC)