Merck
  • Efficient synthesis of oligonucleotide conjugates on solid-support using an (aminoethoxycarbonyl)aminohexyl group for 5'-terminal modification.
Efficient synthesis of oligonucleotide conjugates on solid-support using an (aminoethoxycarbonyl)aminohexyl group for 5'-terminal modification.
Bioorganic & medicinal chemistry letters (2009-03-24)
Naoshi Kojima, Toshie Takebayashi, Akiko Mikami, Eiko Ohtsuka, Yasuo Komatsu
ABSTRACT
Solid-support conjugation at the 5'-terminal primary amine of oligonucleotides is a convenient and powerful method for introducing various functional groups. However, conventional aliphatic amines do not necessarily provide conjugates with sufficient yields. To improve the modification efficacy, we used the amino-linker (aminoethoxycarbonyl)aminohexyl group (ssH-linker), for solid-support conjugation. In the ssH-linker terminal modification, reactive free amino group could be easily presented onto a solid-support due to rapid removal of the amino-protecting group, and activated amino acids or cholesterol molecules could be covalently connected more efficiently than to typical 6-aminohexyl-linkers. Based on these results, the ssH-linker can be a useful terminal modification for the solid-support conjugation of functional molecules.
Sigma-Aldrich
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, ≥93% (AT)
Sigma-Aldrich
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, BioReagent, suitable for plant cell culture, powder
Supelco
Aminoethoxyvinyl glycine hydrochloride, PESTANAL®, analytical standard