Merck
  • Copolymerization of divinylsilyl-11-silicotungstic acid with butyl acrylate and hexanediol diacrylate: synthesis of a highly proton-conductive membrane for fuel-cell applications.
Copolymerization of divinylsilyl-11-silicotungstic acid with butyl acrylate and hexanediol diacrylate: synthesis of a highly proton-conductive membrane for fuel-cell applications.
ChemSusChem (2009-01-27)
James L Horan, Anitha Genupur, Hui Ren, Benjamin J Sikora, Mei-Chen Kuo, Fanqin Meng, Steven F Dec, Gregory M Haugen, Michael A Yandrasits, Steven J Hamrock, Matthew H Frey, Andrew M Herring
ABSTRACT
Highly conducive to high conductivity: Polyoxometalates were incorporated in the backbone of a hydrocarbon polymer to produce proton-conducting films. These first-generation materials contain large, dispersed clusters of polyoxometalates. Although the morphology of these films is not yet optimal, they already demonstrate practical proton conductivities and proton diffusion within the clusters appears to be very high.
Sigma-Aldrich
1,6-Hexanediol diacrylate, technical grade, 80%