Merck
  • Development of a new C14 monolithic silica column containing embedded polar groups for pressurized capillary electrochromatography.
Development of a new C14 monolithic silica column containing embedded polar groups for pressurized capillary electrochromatography.
Journal of separation science (2007-10-11)
Fanggui Ye, Shuting Li, Shulin Zhao
ABSTRACT
Monolithic columns have been prepared with a novel bonded silica stationary phase, tetradecylamine bonded silica (TDAS), and used in pressurized capillary electrochromatography (pCEC). The monolithic silica column matrix was prepared by a sol-gel process and then chemically modified with the spacer (3-glycidoxypropyl)trimethoxysilane and tetradecylamine. The introduced embedded polar amine groups dominated the charge on the surface of the monolithic stationary phase and generated an EOF from cathode to anode under acidic conditions. The tetradecyl hydrophobic chains in TDAS provide chromatographic interactions. The chromatographic characteristics of the prepared monolithic column were studied. Some aromatic compounds including alkylbenzenes, aromatic hydrocarbons, phenols, and anilines were successfully separated on the TDAS monolithic column in pCEC mode. As expected, the TDAS monolithic stationary phases exhibit typical reversed-phase electrochromatographic behavior toward neutral solutes due to the introduced tetradecyl groups. Hydrophobic as well as electrophoretic migration processes within the monoliths were observed in the separation of basic anilines. Symmetrical peaks can be obtained for anilines because the embedded polar amine groups on the surface can effectively shield the adsorption of positively charged analytes onto the stationary phase.
Sigma-Aldrich
Tetradecylamine, 95%