Merck
  • Environmental metabolite, 1,2-diacetylbenzene, produces cytotoxicity through ROS generation in HUVEC cells.
Environmental metabolite, 1,2-diacetylbenzene, produces cytotoxicity through ROS generation in HUVEC cells.
Journal of toxicology and environmental health. Part A (2007-07-27)
Min Kyeong Kim, Kwang Seok Kim, Jae Heun Chung, Jung Hye Kim, Jae-Ryong Kim, Hae Young Chung, Min-Sun Kim
ABSTRACT
Organic solvents are ubiquitous in industrial and household surroundings, and thus individuals are easily exposed. 1,2-Diethylbenzene (DEB) is one of organic solvents contained in gasoline or jet fuels. DEB is absorbed by dermal or inhalation routes, metabolized by cytochrome P-450 in the liver, and ultimately affects mammalian functions. 1,2-Diacetylbenzene (1,2-DAB), which is a putative metabolite of 1,2-DEB, resulted in neuropathological effects on rodent central and peripheral nervous systems. To elucidate the possibility of 1,2-DAB effects on the vascular system, studies were undertaken to examine whether 1,2-DAB induces endothelial cytotoxicity through reactive oxygen species (ROS) generation. Incubation of human umbilical vein endothelial cells (HUVEC) with lower concentrations (4 or 8 microM) of 1,2-DAB induced inhibition of cellular growth and at higher amounts (16 or 32 microM) produced apoptosis. Endothelial cells cultured with 1,2-DAB also showed increased intracellular ROS production and morphological alterations indicative of senescence. Pretreatment with the well-known antioxidant glutathione or N-acetylcysteine (NAC) reduced cytotoxicity induced by 1,2-DAB. Taken together, the results provide evidence that cytotoxicity induced by 1,2-DAB in endothelial cells may be mediated by ROS generation.
Sigma-Aldrich
1,2-Diacetylbenzene, 99%