Merck
  • The non-genotoxicity to rodents of the potent rodent bladder carcinogens o-anisidine and p-cresidine.
The non-genotoxicity to rodents of the potent rodent bladder carcinogens o-anisidine and p-cresidine.
Mutation research (1991-09-01)
J Ashby, P A Lefevre, H Tinwell, G Brunborg, P Schmezer, B Pool-Zobel, R Shanu-Wilson, J A Holme, E J Soderlund, D Gulati
ABSTRACT
The two potent rodent bladder carcinogens o-anisidine and p-cresidine, and the structurally related non-carcinogen 2,4-dimethoxyaniline, have been extensively evaluated for genotoxicity to rodents and found to be inactive. Most data were generated on o-anisidine, an agent that is also only marginally genotoxic in vitro. The two carcinogens induced methaemoglobinaemia in rodents indicating that the chemicals are absorbed and metabolically oxidized. Despite their total lack of genotoxicity in vivo, the two carcinogens have the hall-marks of being genotoxic carcinogens given that most test animals of both sexes of B6C3F1 mice and F344 rats are reported to have succumbed rapidly to malignant bladder cancer. No reasons for this dramatic conflict of test data are so far apparent. The experiments described involve, in one or other combination, 2 strains of mice (including B6C3F1) and 4 strains of rat (including F344), the use of oral and i.p. routes of exposure and observations made after 1, 3 or 6 doses of test chemical. 6 tissues (including the rat bladder) were assayed using 3 genetic endpoints (unscheduled DNA synthesis, DNA single-strand breaks and micronuclei induction). Aroclor-induced rats were employed in one set of experiments with o-anisidine. In the case of one set of mouse bone-marrow micronucleus experiments the same batch of the 3 chemicals as used in the cancer bioassays, and the same strain of mouse, were used. Possible further experiments and the implications of these findings are discussed.
Sigma-Aldrich
2-Methoxy-5-methylaniline, 99%