Merck
  • Monoglycosyl, diglycosyl, and dinucleoside methylenediphosphonates: direct synthesis and antiviral activity.
Monoglycosyl, diglycosyl, and dinucleoside methylenediphosphonates: direct synthesis and antiviral activity.
Carbohydrate research (2006-04-19)
Claude Grison, Stéphane Joliez, E De Clercq, Philippe Coutrot
ABSTRACT
A direct and general access to D-glycosyl 3-, 5-, or 6-methylenediphosphonates, di-D-glycosyl 1,5-, 3,5-, 3,6-, 5,5-, or 6,6-methylenediphosphonates and dithymidine 3',5'-methylenediphosphonate is described. The method involves the one-pot alkylidenediphosphorylation of glycosyl or thymidine derivatives. No antiviral activity was detected against a panel of RNA and DNA viruses.
Product Description
Medronic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylenediphosphonic acid, for molecular studies & mass spectrometry
Sigma-Aldrich
Methylenediphosphonic acid, ≥99%