Merck
[Complexation of cyanidin with cadmium ions in solution].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2005-12-13)
V S Fedenko, S A Shemet, V S Struzhko
ABSTRACT
The effect of complexation between cyanidin (extracted from maize seedling roots) and cadmium ions was established in solution on the basis of absorption spectroscopy and colorimetry data. Spectral parameters for identification of the pigment added to the metal has been established.