Merck
  • Regioselective synthesis of trisubstituted 2,3-dihydrofurans from donor-acceptor cyclopropanes or from reaction of the Corey ylide with alpha-sulfenyl-, alpha-sulfinyl-, or alpha-sulfonylenones.
Regioselective synthesis of trisubstituted 2,3-dihydrofurans from donor-acceptor cyclopropanes or from reaction of the Corey ylide with alpha-sulfenyl-, alpha-sulfinyl-, or alpha-sulfonylenones.
Organic letters (2005-10-08)
Angela M Bernard, Angelo Frongia, Pier P Piras, Francesco Secci, Marco Spiga
ABSTRACT
[reactions: see text] Regioselective synthesis of 2,4,5- or 3,4,5-trisubstituted 2,3-dihydrofurans has been realized by using donor-acceptor cyclopropanes or by a Corey ylide reaction with alpha-sulfenyl-, alpha-sulfinyl-, or alpha-sulfonylenones. The method allowed a straightforward synthesis of the natural product calyxolane B.
Sigma-Aldrich
2,3-Dihydrofuran, 99%