Merck
  • Unusually high phosphodiesterolytic activity of La(III) hydroxide complexes stabilized by glycine derivatives.
Unusually high phosphodiesterolytic activity of La(III) hydroxide complexes stabilized by glycine derivatives.
Chemical communications (Cambridge, England) (2003-08-23)
Felipe Medrano, Antonio Calderón, Anatoly K Yatsimirsky
ABSTRACT
Glycine and N,N-dimethylglycine stabilize La(III) hydroxide complexes of the type La2L2(OH)4 which possess phosphodiesterolytic activity close to that observed with most active tetravalent cations like Ce(IV).
Sigma-Aldrich
Lanthanum(III) hydroxide, 99.9% trace metals basis