Merck
  • Thiation of 2'-deoxy-5,6-dihydropyrimidine nucleosides with Lawesson's reagent: characterisation of oxathiaphosphepane intermediates.
Thiation of 2'-deoxy-5,6-dihydropyrimidine nucleosides with Lawesson's reagent: characterisation of oxathiaphosphepane intermediates.
Chemical communications (Cambridge, England) (2003-04-22)
Frédéric Peyrane, Jean-Louis Fourrey, Pascale Clivio
ABSTRACT
Treatment of 2'-deoxy-3',5'-dithexyldimethylsilyl-5,6-dihydrouridine with Lawesson's reagent led to the expected C4-thiolated derivative together with a number of oxathiaphosphepane isomers which resulted from the heat reversible incorporation of an AnPS2 unit within the 2'-deoxyribose moiety explaining the subsequent anomerisation of the 5,6-dihydropyrimidine nucleosides.
Sigma-Aldrich
Lawesson reagent, 97%