Merck
  • Taurohyodeoxycholate- and tauroursodeoxycholate-induced hypercholeresis is augmented in bile duct ligated rats.
Taurohyodeoxycholate- and tauroursodeoxycholate-induced hypercholeresis is augmented in bile duct ligated rats.
Journal of hepatology (2003-01-28)
Leonardo Baiocchi, Gianfranco Alpini, Shannon Glaser, Mario Angelico, Domenico Alvaro, Heather Francis, Marco Marzioni, Jo Lynne Phinizy, Barbara Barbaro, Gene LeSage
ABSTRACT
Taurohyodeoxycholate (THDCA) and tauroursodeoxycholate (TUDCA) induce more bile flow per molecule excreted compared to endogenous bile acids. The aim of this study is to determine if the hypercholeretic effect of tauroursodeoxycholate or taurohyodeoxycholate in normal and bile duct ligated (BDL) rats is due to increased ductal secretion. Normal or BDL rats were infused with tauroursodeoxycholate or taurohyodeoxycholate and bile flow, bicarbonate, bile salt, cholesterol, and phospholipid secretion were measured. Cholangiocytes were stimulated with taurohyodeoxycholate or tauroursodeoxycholate, and secretin-stimulated secretion was measured. Taurohyodeoxycholate and tauroursodeoxycholate increased bile flow more in BDL than normal rats. Tauroursodeoxycholate increased bicarbonate secretion more in BDL compared to normal rats. Taurohyodeoxycholate when infused with taurocholate increased bile flow (but not phospholipid excretion) to a greater degree in BDL compared to normal rats. Taurohyodeoxycholate and tauroursodeoxycholate decreased secretin-stimulated cholangiocyte secretion. Consistent with a ductal origin for bile acid-induced hypercholeresis, taurohyodeoxycholate and tauroursodeoxycholate produced a greater hypercholeresis in BDL than normal rats. Tauroursodeoxycholate- (but not taurohyodeoxycholate-) stimulated hypercholeresis is associated with increased HCO(3)(-) secretion. Tauroursodeoxycholate increases biliary HCO(3)(-) secretion by a mechanism unrelated to secretin-stimulated cholangiocyte secretion. Taurohyodeoxycholate-induced hypercholeresis in BDL rats is unrelated to enhanced phospholipid excretion.
Sigma-Aldrich
Sodium taurohyodeoxycholate hydrate, ≥98%