Merck
Dealkylation of phosphonate esters with chlorotrimethylsilane.
Nucleosides, nucleotides & nucleic acids (2001-09-21)
A J Gutierrez, E J Prisbe, J C Rohloff
ABSTRACT
Chlorotrimethylsilane completely dealkylates phosphonate esters at elevated temperature in a sealed reaction vessel. These conditions are tolerated by a variety of functional groups and lead to high conversions of dimethyl, diethyl and diisopropyl phosphonates to their corresponding phosphonic acids.
Sigma-Aldrich
Dimethyl phosphite, 98%