Merck
  • Modulation of glutamate exocytosis by redox changes of superficial thiol groups in rat cerebrocortical synaptosomes.
Modulation of glutamate exocytosis by redox changes of superficial thiol groups in rat cerebrocortical synaptosomes.
Neuroscience letters (1999-12-20)
F Zoccarato, L Cavallini, M Valente, A Alexandre
ABSTRACT
The treatment of cerebral cortex synaptosomes with the membrane impermeable thiol reagent 5,5'-dithio-bis-(2-nitrobenzoic acid) (DTNB) induces a long-lasting partial inhibition (about 40%) of the KCl-stimulated Ca2+-dependent exocytosis of glutamate. Synaptosomes are not damaged by the treatment. The increase of cytoplasmic free Ca2+ concentration ([Ca2+]i) upon depolarization is not affected by DTNB. The inhibition is observed also if exocytosis is induced with the Ca2+-ionophore ionomycin. In all cases the inhibition is reversed by the impermeable reductant glutathione (GSH). Similarly the inhibition of exocytosis by H2O2 (Zoccarato, F., Valente, M. and Alexandre, A., Hydrogen peroxide induces a long-lasting inhibition of the Ca2+-dependent glutamate release in cerebrocortical synaptosomes without interfering with cytosolic Ca2+. J. Neurochem., 64 (1995) 2552-2558.) is reversed by GSH. It is concluded that redox changes (possibly thiol-disulfide transitions) of superficial groups modulate the exocytotic apparatus directly. In an attempt to identify the protein(s) involved in this novel type of control, we evidenced DTNB (H2O2) reactive bands at 35 and at 85-150 kDa which can be labeled with a monobromotrimethylammoniobimane bromide (qBBr) derivatization.
Sigma-Aldrich
Monobromo(trimethylammonio)bimane bromide, suitable for fluorescence, ≥90.0% (HPLC)