- Role of reactive oxygen species and Ca(2+) dissociation from the myofilaments in determination of Ca(2+) wave propagation in rat cardiac muscle.
Role of reactive oxygen species and Ca(2+) dissociation from the myofilaments in determination of Ca(2+) wave propagation in rat cardiac muscle.
Ca(2+) waves are initiated not only by Ca(2+) leak from the sarcoplasmic reticulum (SR), but also by Ca(2+) dissociation from the myofilaments in the myocardium with nonuniform contraction. We investigated whether contractile properties and the production of reactive oxygen species (ROS) affect Ca(2+) wave propagation. Trabeculae were obtained from 76 rat hearts. Force was measured with a strain gauge, sarcomere length with a laser diffraction technique, and [Ca(2+)](i) with fura-2 and a CCD camera (24°C, 2.0mmol/L [Ca(2+)](o)). ROS production was estimated from 2',7'-dichlorofluorescein (DCF) fluorescence. Trabeculae were regionally exposed to a jet of solution containing 1) 10mmol/L Ca(2+) to initiate Ca(2+) waves by SR Ca(2+) leak due to Ca(2+) overload within the jet-exposed region, and 2) 0.2mmol/L Ca(2+) or 5mmol/L caffeine to initiate such waves by Ca(2+) dissociation from the myofilaments due to nonuniform contraction. Ca(2+) waves were induced by stimulus trains for 7.5s. Ten-percent muscle stretch increased DCF fluorescence and accelerated Ca(2+) waves initiated due to both Ca(2+) overload and nonuniform contraction. Preincubation with 3μmol/L diphenyleneiodonium or 10μmol/L colchicine suppressed the increase in DCF fluorescence but suppressed acceleration of Ca(2+) waves initiated only due to Ca(2+) overload. Irrespective of preincubation with colchicine, reduction of force after the addition of 10μmol/L blebbistatin did not decelerate Ca(2+) waves initiated due to Ca(2+) overload, while it did decelerate waves initiated due to nonuniform contraction. These results suggest that Ca(2+) wave propagation is modulated by ROS production through an intact microtubule network only during stretch and may be additionally modulated by Ca(2+) dissociated from the myofilaments in the case of nonuniform contraction.