Merck
  • Cloning and functional expression of a vascular smooth muscle endothelin 1 receptor.
Cloning and functional expression of a vascular smooth muscle endothelin 1 receptor.
Proceedings of the National Academy of Sciences of the United States of America (1991-04-15)
H Y Lin, E H Kaji, G K Winkel, H E Ives, H F Lodish
ABSTRACT
By screening a cDNA library derived from the A10 rat vascular smooth muscle cell line for functional expression in COS cells, we have isolated a high-affinity receptor for endothelin 1 (Kd = 476 pM) and endothelin 2. The affinity of the cloned endothelin receptor for endothelin 3 is greater than 100 times less in A10 cells and in a CHO cell line stably transformed by the endothelin receptor cDNA. The 426-amino acid receptor polypeptide has seven putative hydrophobic transmembrane domains and is presumed to be a member of the family of guanine nucleotide-binding regulatory (G) protein-coupled receptors. Microinjection of in vitro transcripts of the cloned cDNA into CHO cells confers a transient increase in intracellular calcium in response to endothelin 1, indicating that the receptor is functional and couples to the appropriate G protein(s). RNA analysis reveals high expression in rat lung and heart, tissues known to exhibit binding to iodinated endothelin 1.
Product Description
A10-85, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.