Merck
  • SMYD4 monomethylates PRMT5 and forms a positive feedback loop to promote hepatocellular carcinoma progression.
SMYD4 monomethylates PRMT5 and forms a positive feedback loop to promote hepatocellular carcinoma progression.
Cancer science (2024-03-05)
Zhenyu Zhou, Zheng Chen, Qianlei Zhou, Shiyu Meng, Juanyi Shi, Sintim Mui, Hai Jiang, Jianhong Lin, Gui He, Wenbin Li, Jianlong Zhang, Jie Wang, Chuanchao He, Yongcong Yan, Zhiyu Xiao
ABSTRACT
Both lysine and arginine methyltransferases are thought to be promising therapeutic targets for malignant tumors, yet how these methyltransferases function in malignant tumors, especially hepatocellular carcinoma (HCC), has not been fully elucidated. Here, we reported that SMYD4, a lysine methyltransferase, acts as an oncogene in HCC. SMYD4 was highly upregulated in HCC and promoted HCC cell proliferation and metastasis. Mechanistically, PRMT5, a well-known arginine methyltransferase, was identified as a SMYD4-binding protein. SMYD4 monomethylated PRMT5 and enhanced the interaction between PRMT5 and MEP50, thereby promoting the symmetrical dimethylation of H3R2 and H4R3 on the PRMT5 target gene promoter and subsequently activating DVL3 expression and inhibiting expression of E-cadherin, RBL2, and miR-29b-1-5p. Moreover, miR-29b-1-5p was found to inversely regulate SMYD4 expression in HCC cells, thus forming a positive feedback loop. Furthermore, we found that the oncogenic effect of SMYD4 could be effectively suppressed by PRMT5 inhibitor in vitro and in vivo. Clinically, high coexpression of SMYD4 and PRMT5 was associated with poor prognosis of HCC patients. In summary, our study provides a model of crosstalk between lysine and arginine methyltransferases in HCC and highlights the SMYD4-PRMT5 axis as a potential therapeutic target for the treatment of HCC.
Sigma-Aldrich
ChIPAb+ Dimethyl-Histone H4 (Arg3) Symmetric - ChIP Validated Antibody and Primer Set, from rabbit, purified by affinity chromatography