Merck
  • Gas chromatographic separation of methoxylated polychlorinated biphenyl atropisomers.
Gas chromatographic separation of methoxylated polychlorinated biphenyl atropisomers.
Journal of chromatography. A (2008-09-02)
Izabela Kania-Korwel, Sandhya M Vyas, Yang Song, Hans-Joachim Lehmler
ABSTRACT
Several polychlorinated biphenyls (PCBs) and their hydroxylated metabolites display axial chirality. Here we describe an enantioselective, gas chromatographic separation of methylated derivatives of hydroxylated (OH-)PCB atropisomers (MeO-PCB) using a chemically bonded beta-cyclodextrin column (Chirasil-Dex). The atropisomers of several MeO-PCBs could be separated on this column with resolutions ranging from 0.42 to 0.87 under isothermal or temperature-programmed conditions. In addition, the enantiomeric fraction of OH-PCB 136 metabolites was determined in male and female rats treated with racemic PCB 136. The methylated derivatives of two OH-PCB 136 metabolites showed an enantiomeric enrichment in liver tissue, whereas PCB 136 itself was near racemic.
Sigma-Aldrich
N-Methyl-N-nitroso-p-toluenesulfonamide, purum, ≥98.0% (HPLC)