Merck
  • High-Throughput CD36 Phenotyping on Human Platelets Based on Sandwich ELISA and Mutant Gene Analysis.
High-Throughput CD36 Phenotyping on Human Platelets Based on Sandwich ELISA and Mutant Gene Analysis.
Transfusion medicine and hemotherapy : offizielles Organ der Deutschen Gesellschaft fur Transfusionsmedizin und Immunhamatologie (2024-02-05)
Honghong He, Longhai Tang, Yiming Jin, Yujue Wang, Hongmei Wang, Shaohua Ding, Yezhou Chen, Jingjing Tian, Mingyuan Wang, Shengbao Duan
ABSTRACT
CD36 deficiency is closely associated with fetal/neonatal alloimmune thrombocytopenia, platelet transfusion refractoriness, and other hemorrhage disorders, particularly in Asian and African populations. There is a clinical need for rapid and high-throughput methods of platelet CD36 (pCD36) phenotyping to improve the availability of CD36 typing of donors and assist clinical blood transfusions for patients with anti-CD36 antibodies. Such methods can also support the establishment of databases of pCD36-negative phenotypes. A sandwich enzyme-linked immunosorbent assay (ELISA) for CD36 phenotyping of human platelets was developed using anti-CD36 monoclonal antibodies. The reliability of the assay was evaluated by calculating the intra-assay and inter-assay coefficients of variation (CV). A total of 1,691 anticoagulant whole blood samples from healthy blood donors were randomly selected. PCD36 expression was measured using a sandwich ELISA. PCD36 deficiency was confirmed by flow cytometry (FC). Mutations underlying pCD36 deficiency were identified using polymerase chain reaction sequence-based typing (PCR-SBT). The sandwich ELISA for pCD36 phenotyping had high reliability (intra-assay CV, 2.1-4.8%; inter-assay CV, 2.3-5.2%). The sandwich ELISA was used to screen for CD36 expression on platelets isolated from 1,691 healthy blood donors. Of these, 36 samples were pCD36-negative. FC demonstrated absence of CD36 expression on monocytes in three of the 36 cases. In the present study population, the frequency of CD36 deficiency was 2.13% (36/1,691), of which 0.18% (3/1,691) was type I deficiency and 1.95% (33/1,691) was type II deficiency. In addition, we used PCR-SBT to characterize the gene mutations in exons 3-14 of the CD36 gene in 27 cases of CD36 deficiency and discovered 10 types of mutations in 13 pCD36-negative samples. The present study describes the development and characterization of a highly reliable sandwich ELISA for high-throughput screening for pCD36 expression. This novel method is feasible for clinical applications and provides a useful tool for the establishment of databases of pCD36-negative phenotype donors.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD36 antibody produced in mouse, clone TR9, purified immunoglobulin, buffered aqueous solution