Merck
  • LRIT3 expression in cone photoreceptors restores post-synaptic bipolar cell signalplex assembly and partial function in Lrit3 -/- mice.
LRIT3 expression in cone photoreceptors restores post-synaptic bipolar cell signalplex assembly and partial function in Lrit3 -/- mice.
iScience (2023-04-24)
Ronald G Gregg, Nazarul Hasan, Bart G Borghuis
ABSTRACT
Complete congenital stationary night blindness (cCSNB) is a heterogeneous disorder characterized by poor dim-light vision, myopia, and nystagmus that is caused by mutations in genes critical for signal transmission between photoreceptors and depolarizing bipolar cells (DBCs). One such gene, LRIT3, is required for assembly of the post-synaptic signaling complex (signalplex) at the dendritic tips of DBCs, although the number of signalplex components impacted is greater in cone DBCs (all components) than in rod bipolar cells (only TRPM1 and Nyctalopin). Here we show that rAAV-mediated expression of LRIT3 in cones results in robust rescue of cone DBC signalplex components and partially restores downstream visual function, as measured by the light-adapted electroretinogram (ERG) b-wave and electrophysiological recordings of bipolar cells (BCs) and RGCs. These data show that LRIT3 successfully restores partial function to cone DBCs most likely in a trans-synaptic manner, potentially paving the way for therapeutic intervention in LRIT3-associated cCSNB.
Sigma-Aldrich
Donkey Anti-Guinea Pig IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey