Merck
  • Stimulation of Akt Phosphorylation and Glucose Transport by Metalloporphyrins with Peroxynitrite Decomposition Catalytic Activity.
Stimulation of Akt Phosphorylation and Glucose Transport by Metalloporphyrins with Peroxynitrite Decomposition Catalytic Activity.
Catalysts (Basel, Switzerland) (2023-05-01)
Amanda M Eccardt, Ross J Pelzel, Thomas P Bell, Jonathan S Fisher
ABSTRACT
Iron porphyrin molecules such as hemin and iron(III) 4,4',4″,4‴-(porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid) (FeTBAP) have previously been shown to influence insulin signaling and glucose metabolism. We undertook this study to determine whether a catalytic action of iron porphyrin compounds would be related to their stimulation of insulin signaling and glucose uptake in C2C12 myotubes. FeTBAP did not display nitrite reductase activity or alter protein S-nitrosylation in myotubes, eliminating this as a candidate mode by which FeTBAP could act. FeTBAP displayed peroxynitrite decomposition catalytic activity in vitro. Additionally, in myotubes FeTBAP decreased protein nitration. The peroxynitrite decomposition catalyst Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride (FeTPPS) also decreased protein nitration in myotubes, but the iron porphyrin Fe(III)tetrakis(1-methyl-4-pyridyl)porphyrin pentachlorideporphyrin pentachloride (FeTMPyP) did not. FeTBAP and FeTPPS, but not FeTMPyP, showed in vitro peroxidase activity. Further, FeTBAP and FeTPPs, but not FeTMPyP, increased Akt phosphorylation and stimulated glucose uptake in myotubes. These findings suggest that iron porphyrin compounds with both peroxynitrite decomposition activity and peroxidase activity can stimulate insulin signaling and glucose transport in skeletal muscle cells.