Merck
  • The Mobilome: Metagenomic Analysis of Circular Plasmids, Viruses, and Other Extrachromosomal Elements.
The Mobilome: Metagenomic Analysis of Circular Plasmids, Viruses, and Other Extrachromosomal Elements.
Methods in molecular biology (Clifton, N.J.) (2019-10-05)
Patrick Denis Browne, Witold Kot, Tue Sparholt Jørgensen, Lars Hestbjerg Hansen
ABSTRACT
Isolation, sequencing, and analysis of circular genetic elements bring new insights to mobile genetic elements related to microbial ecology. One method used to study circular plasmids, viruses, and other elements is called the mobilome method. The mobilome method presented here is an unamplified mobilome approach allowing fast isolation of circular DNA elements from a variety of samples followed by directly building unamplified Illumina-compatible sequencing libraries using enzymatic tagging and fragmentation. Several methods for bioinformatic analysis of mobilome data are also suggested.
Sigma-Aldrich
Bovine Serum Albumin solution, 20 mg/mL in H2O, low bioburden, protease free
Sigma-Aldrich
MetaPolyzyme, lyophilized powder