Merck
  • ARHGEF40 promotes non-small cell lung cancer proliferation and invasion via the AKT-Wnt axis by binding to RhoA.
ARHGEF40 promotes non-small cell lung cancer proliferation and invasion via the AKT-Wnt axis by binding to RhoA.
Molecular carcinogenesis (2022-08-25)
Jian Gu, Xiupeng Zhang, Guiyang Jiang, Qingchang Li, Enhua Wang, Juanhan Yu
ABSTRACT
Rho guanine nucleotide exchange factor 40 (ARHGEF40) is a member of the Dbl-family of guanine nucleotide factor proteins. However, its expression pattern and biological function in malignant tumors, notably in nonsmall cell lung cancer (NSCLC) are currently unknown. The present study demonstrated that ARHGEF40 was highly expressed in NSCLC specimens and that its expression was significantly associated with advanced TNM stage (p < 0.001), lymph node metastasis (p = 0.002), and poor prognosis (p = 0.0056). In addition, ARHGEF40 accelerated nuclear translocation of the key component β-catenin and increased the expression levels of the Wnt signaling pathway targets c-myc, cyclin D1 and MMP7. Moreover, it promoted lung cancer cell proliferation and invasion in vitro and in vivo. To elucidate the underlying molecular mechanism, the current study demonstrated that ARHGEF40 could induce activation of the Wnt signaling pathway by increasing the phosphorylation levels of AKT and GSK3β via interaction with RhoA. Moreover, the Dbl homology (DH)-pleckstrin homology (PH) domain of ARHGEF40 was responsible for this interaction. Its deletion abolished the binding, which blocked the activation of the Wnt signaling. Taken together, the data indicated that ARHGEF40 promoted the malignant phenotype of lung cancer cells by activating the AKT-Wnt axis. This was achieved by its interaction with RhoA via the DH-PH domain. ARHGEF40 may serve as a novel target for NSCLC treatment.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARHGEF40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1