Merck
  • Cold-inducible RNA-binding protein regulates cyclin B1 against spermatogenesis arrest caused by heat stress.
Cold-inducible RNA-binding protein regulates cyclin B1 against spermatogenesis arrest caused by heat stress.
Andrology (2021-10-11)
Heyu Liu, Chengcheng Xu, Meng Bao, Jin Huang, Liping Zou, Xiaorong Fan, Changhong Zhu, Wei Xia
ABSTRACT
Spermatogenesis arrest and spermatogenic cell apoptosis occur in the testes of heat-stressed mice. Although heat stress-induced spermatogenic cell apoptosis is due to the decreased expression of cold-inducible RNA-binding protein (CIRBP), it remains unclear whether spermatogenesis arrest is also affected by CIRBP. Additionally, the specific mechanism by which CIRBP regulates spermatogenic cell apoptosis or inhibits spermatogenesis remains to be elucidated. To investigate the mechanism by which CIRBP contributes to heat stress-induced testicular spermatogenesis arrest. Target mRNAs downstream of CIRBP in testicular tissue of BALB/c mice, exposed or not to heat stress, were sequenced. Sequencing data were subjected to bioinformatics analysis to identify key mRNAs and pathways associated with heat stress-induced spermatogenic damage. The link between CIRBP and its target mRNA Ccnb1 (cyclin B1) was verified by western blotting, flow cytometry, and RNA pulldown assays, and the ability of CIRBP to inhibit germ cell cycle arrest by regulating cyclin B1 expression was investigated in a mouse spermatocyte cell line (GC-2spd). Changes in mRNA expression downstream of CIRBP were mainly associated with the cell cycle and RNA binding, transport and splicing. Cyclin B1 was found to regulate the G2/M transition during the first meiotic division of spermatogenic cells. Further, CIRBP was shown to bind directly to the 3'-untranslated region of Ccnb1 mRNA and was associated with cyclin B1-induced inhibition of spermatogenesis arrest. In conclusion, our results provide strong evidence that CIRBP may exert its key function in heat stress-induced testicular spermatogenic cell injury partly by regulating the expression of Ccnb1, the product of which inhibits spermatogenesis arrest.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, FITC conjugate, Species Adsorbed, Chemicon®, from goat