Merck
XPF activates break-induced telomere synthesis.
Nature communications (2022-10-03)
Chia-Yu Guh, Hong-Jhih Shen, Liv WeiChien Chen, Pei-Chen Chiu, I-Hsin Liao, Chen-Chia Lo, Yunfei Chen, Yu-Hung Hsieh, Ting-Chia Chang, Chien-Ping Yen, Yi-Yun Chen, Tom Wei-Wu Chen, Liuh-Yow Chen, Ching-Shyi Wu, Jean-Marc Egly, Hsueh-Ping Catherine Chu
ABSTRACT
Alternative Lengthening of Telomeres (ALT) utilizes a recombination mechanism and break-induced DNA synthesis to maintain telomere length without telomerase, but it is unclear how cells initiate ALT. TERRA, telomeric repeat-containing RNA, forms RNA:DNA hybrids (R-loops) at ALT telomeres. We show that depleting TERRA using an RNA-targeting Cas9 system reduces ALT-associated PML bodies, telomere clustering, and telomere lengthening. TERRA interactome reveals that TERRA interacts with an extensive subset of DNA repair proteins in ALT cells. One of TERRA interacting proteins, the endonuclease XPF, is highly enriched at ALT telomeres and recruited by telomeric R-loops to induce DNA damage response (DDR) independent of CSB and SLX4, and thus triggers break-induced telomere synthesis and lengthening. The attraction of BRCA1 and RAD51 at telomeres requires XPF in FANCM-deficient cells that accumulate telomeric R-loops. Our results suggest that telomeric R-loops activate DDR via XPF to promote homologous recombination and telomere replication to drive ALT.
Sigma-Aldrich
Anti-FANCM Antibody, clone CV5.1, clone CV5.1, 0.5 mg/mL, from mouse
Sigma-Aldrich
Anti-DNA-RNA Hybrid Antibody, clone S9.6, clone S9.6, from mouse