Merck
  • Preparation of hollow spheres with controllable interior structures by heterogeneous contraction.
Preparation of hollow spheres with controllable interior structures by heterogeneous contraction.
Chemical communications (Cambridge, England) (2010-08-18)
Jianguo Guan, Fangzhi Mou, Zhigang Sun, Weidong Shi
ABSTRACT
We present a simple and effective heterogeneous contraction method to fabricate hollow spheres with controllable interior structures (ranging from solid, simple hollow to core-in-hollow-wall, double-wall hollow and core-in-double-hollow-wall spheres) by a non-equilibrium heat-treatment process of gel precursors with a high heating rate.
Sigma-Aldrich
Iron(III) citrate, technical grade