Merck
  • Discovery of a novel family of CDK inhibitors with the program LIDAEUS: structural basis for ligand-induced disordering of the activation loop.
Discovery of a novel family of CDK inhibitors with the program LIDAEUS: structural basis for ligand-induced disordering of the activation loop.
Structure (London, England : 1993) (2003-04-08)
Su Ying Wu, Iain McNae, George Kontopidis, Steven J McClue, Campbell McInnes, Kevin J Stewart, Shudong Wang, Daniella I Zheleva, Howard Marriage, David P Lane, Paul Taylor, Peter M Fischer, Malcolm D Walkinshaw
ABSTRACT
A family of 4-heteroaryl-2-amino-pyrimidine CDK2 inhibitor lead compounds was discovered with the new database-mining program LIDAEUS through in silico screening. Four compounds with IC(50) values ranging from 17 to 0.9 microM were selected for X-ray crystal analysis. Two distinct binding modes are observed, one of which resembles the hydrogen bonding pattern of bound ATP. In the second binding mode, the ligands trigger a conformational change in the activation T loop by inducing movement of Lys(33) and Asp(145) side chains. The family of molecules discovered provides an excellent starting point for the design and synthesis of tight binding inhibitors, which may lead to a new class of antiproliferative drugs.
Sigma-Aldrich
Adenosine 5′-triphosphate–Agarose, lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution