Merck
  • Dissociation and inDrops microfluidic encapsulation of human gut tissues for single-cell atlasing studies.
Dissociation and inDrops microfluidic encapsulation of human gut tissues for single-cell atlasing studies.
STAR protocols (2022-07-27)
Alan J Simmons, Ken S Lau
ABSTRACT
In droplet-based single-cell RNA-sequencing (scRNA-seq) experiments, cells, along with some of their surrounding buffer and ambient material, are encapsulated into droplets for mRNA capture and barcoding. This protocol details the steps for human gut tissue dissociation using cold active protease, and subsequent isolation of single epithelial cells, with enrichment of viability through washes. Next, the steps for encapsulation on the inDrops scRNA-seq platform are described. This procedure has been demonstrated to be applicable to polyps, cancers, and inflamed tissues. For complete details on the use and execution of this protocol, please refer to Chen et al. (2021).
Sigma-Aldrich
Albumin, Bovine Serum, 10% Aqueous Solution, Nuclease-Free