Merck
  • Mocetinostat suppresses epidural fibrosis following laminectomy by inhibiting myofibroblast activation and increasing apoptosis.
Mocetinostat suppresses epidural fibrosis following laminectomy by inhibiting myofibroblast activation and increasing apoptosis.
European review for medical and pharmacological sciences (2021-05-19)
W-J Wu, J Wang, J Liang, Q Zhou, Y Liang
ABSTRACT
The article "Mocetinostat suppresses epidural fibrosis following laminectomy by inhibiting myofibroblast activation and increasing apoptosis, by W.-J. Wu, J. Wang, J. Liang, Q. Zhou, Y. Liang, published in Eur Rev Med Pharmacol Sci 2020; 24 (8): 4467-4475-DOI: 10.26355/eurrev_202004_21029-PMID: 32373984" has been withdrawn from the authors due to some errors in the data. The Publisher apologizes for any inconvenience this may cause. https://www.europeanreview.org/article/21029.
Sigma-Aldrich
MGCD0103, ≥95% (HPLC)