Merck
  • Visible-Light-Induced Synthesis of 1,2,3,4-Tetrahydroquinolines through Formal [4+2] Cycloaddition of Acyclic α,β-Unsaturated Amides and Imides with N,N-Dialkylanilines.
Visible-Light-Induced Synthesis of 1,2,3,4-Tetrahydroquinolines through Formal [4+2] Cycloaddition of Acyclic α,β-Unsaturated Amides and Imides with N,N-Dialkylanilines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-12-11)
Kennosuke Itoh, Shun-Ichi Nagao, Ken Tokunaga, Shigeto Hirayama, Fumika Karaki, Takaaki Mizuguchi, Kenichiro Nagai, Noriko Sato, Mitsuaki Suzuki, Masashi Hashimoto, Hideaki Fujii
ABSTRACT
1,2,3,4-Tetrahydroquinolines should be applicable to the development of new pharmaceutical agents. A facile synthesis of 1,2,3,4-tetrahydroquinolines that is achieved by a photoinduced formal [4+2] cycloaddition reaction of acyclic α,β-unsaturated amides and imides with N,N-dialkylanilines under visible-light irradiation, in which a new IrIII complex photosensitizer, a thiourea, and an oxidant act cooperatively in promoting the reaction, is reported. The photoreaction enables the synthesis of a wide variety of 1,2,3,4-tetrahydroquinolines, while controlling the trans/cis diastereoselectivity (>99:1) and constructing contiguous stereogenic centers. A chemoselective cleavage of an acyclic imide auxiliary is demonstrated.
Sigma-Aldrich
[Ir(df(CF3)ppy)2(4,4′-(OMe)2bpy]BF4, ≥95%