Merck
  • Cyclin-dependent kinase 4/6 inhibitors suppress tumor growth in extramammary Paget's disease.
Cyclin-dependent kinase 4/6 inhibitors suppress tumor growth in extramammary Paget's disease.
Cancer science (2021-12-07)
Shinya Kitamura, Teruki Yanagi, Takuya Maeda, Hideyuki Ujiie
ABSTRACT
Extramammary Paget's disease (EMPD) is a rare adnexal neoplasm commonly seen in the genital areas among the senior population. The prognosis of advanced EMPD is not favorable; thus, the development of potential treatments has long been sought. Cyclin-dependent kinase (CDK) 4/6 inhibitors such as abemaciclib and palbociclib have been proven effective against metastatic breast cancer; however, no studies have addressed CDK4/6 inhibitors as an EMPD treatment. We herein examine the efficacy of CDK4/6 inhibitors against an EMPD patient-derived xenograft (PDX) model. Abemaciclib (50 mg/kg/day) or palbociclib (120 mg/kg/day) was given orally to tumor-bearing NOD/Scid mice over a 3-week period. We also investigated the protein expression levels of CDK4/6 and cyclin D1 through immunohistochemical staining using EMPD clinical samples. Treatment with abemaciclib or palbociclib as a single agent was found to significantly suppress tumor growth in EMPD-PDX. The Ki-67-positive ratio of the treated EMPD-PDX tumors was significantly lower than that of the nontreated tumors. Clinically, the expression levels of CDK4 and cyclin D1 were significantly higher in the EMPD tumor cells than in the normal epidermis. Our results suggest that CDK4/6 inhibitors could be novel and potent therapeutics for the treatment of EMPD.
Sigma-Aldrich
Palbociclib, ≥98% (HPLC)