Merck
  • Diagnostic NH and OH vibrations for oxazolone and diketopiperazine structures: b2 from protonated triglycine.
Diagnostic NH and OH vibrations for oxazolone and diketopiperazine structures: b2 from protonated triglycine.
Journal of the American Society for Mass Spectrometry (2011-09-29)
Da Wang, Kerim Gulyuz, Corey N Stedwell, Nick C Polfer
ABSTRACT
We present infrared multiple photon dissociation (IRMPD) spectra in the hydrogen stretching region of the simplest b fragment, b(2) from protonated triglycine, contrasted to that of protonated cyclo(Gly-Gly). Both spectra confirm the presence of intense, diagnostic vibrations linked to the site of proton attachment. Protonated cyclo(Gly-Gly) serves as a reference spectrum for the diketopiperazine structure, showing a diagnostic O-H(+) stretch of the protonated carbonyl group at 3585 cm(-1). Conversely, b(2) from protonated triglycine exhibits a strong band at 3345 cm(-1), associated with the N-H stretching mode of the protonated oxazolone ring structure. Other weaker N-H stretches can also be discerned, such as the amino NH(2) and amide NH bands. These results demonstrate the usefulness of the hydrogen stretching region, and hence benchtop optical parametric oscillator/amplifier (OPO/A) set-ups, in making structural assignments of product ions in collision-induced dissociation (CID) of peptides.
Sigma-Aldrich
Glycine anhydride, cyclic dipeptide
Sigma-Aldrich
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly-Gly