Merck
Anti-formyl peptide antibodies.
Bioorganic & medicinal chemistry letters (2007-02-13)
Fujie Tanaka, Teresa Jones, Diane Kubitz, Richard A Lerner
ABSTRACT
Antibodies that selectively bind to N-formylmethionyl leucyl phenylalanine (fMLF, also known as fMLP) have been generated. These antibodies bound to fMLF with higher affinity than to non-formylated peptide MLF: the differences in the binding energies between fMLF and MLF were 1.4->2.1 kcal/mol.
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)