Merck
  • A protocol for displaying viral envelope glycoproteins on the surface of vesicular stomatitis viruses.
A protocol for displaying viral envelope glycoproteins on the surface of vesicular stomatitis viruses.
STAR protocols (2020-12-31)
Touraj Aligholipour Farzani, Angela Chov, Alon Herschhorn
ABSTRACT
We describe the production of single-cycle (sc) and replication-competent recombinant vesicular stomatitis viruses (rcVSVs) displaying heterologous envelope glycoproteins (Envs) on their surface. We prepare scVSVs by transiently expressing HIV-1 Envs or SARS-CoV-2 spike followed by infection of the cells with scVSV particles, which do not carry the vsv-g gene. To prepare rcVSVs, we replace the vsv-g with a specific env-encoding gene, transfect cells with multiple plasmids for production of the genomic RNA and viral proteins, and rescue replication-competent viruses.
Sigma-Aldrich
trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate, ACS reagent, for complexometry, 98%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O