Merck
  • MicroRNA-210 Mediates Hypoxia-Induced Repression of Spontaneous Transient Outward Currents in Sheep Uterine Arteries During Gestation.
MicroRNA-210 Mediates Hypoxia-Induced Repression of Spontaneous Transient Outward Currents in Sheep Uterine Arteries During Gestation.
Hypertension (Dallas, Tex. : 1979) (2021-03-02)
Xiang-Qun Hu, Chiranjib Dasgupta, Rui Song, Monica Romero, Sean M Wilson, Lubo Zhang
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-Ryanodine Receptor 2 Antibody, serum, Chemicon®