Merck
  • Interrelation between the metabolism of L-isoleucine and L-allo-isoleucine in patients with maple syrup urine disease.
Interrelation between the metabolism of L-isoleucine and L-allo-isoleucine in patients with maple syrup urine disease.
Pediatric research (1989-01-01)
U Wendel, U Langenbeck, J W Seakins
ABSTRACT
The nonprotein amino acid L-allo-isoleucine is formed endogenously in maple syrup urine disease patients from (R)-3-methyl-2-oxo-pentanoic acid. During strict metabolic balance, the plasma L-allo-isoleucine/L-isoleucine ratio correlates inversely with the residual activity of the branched-chain 2-oxoacid dehydrogenase in fibroblasts and thus constitutes a relevant in vivo parameter of the severity of the metabolic defect in MSUD patients.
Sigma-Aldrich
L-allo-Isoleucine