Merck
  • Synthesis, Molecular Docking and Cytotoxic Activity Evaluation of Organometallic Thiolated Gold(I) Complexes.
Synthesis, Molecular Docking and Cytotoxic Activity Evaluation of Organometallic Thiolated Gold(I) Complexes.
Iranian journal of pharmaceutical research : IJPR (2021-03-09)
Zeinab Faghih, Alireza Yazdani Kachoei, Hossein Alizadeh, Suphia Emamdoost, Shima Shirkhan, Masood Fereidoonnezhad
ABSTRACT
The complex [(PhCH2NC)AuCl], 1, was prepared by the reaction of [(Me2S)AuCl], A, with an equimolar amount of benzyl isocyanide (PhCH2NC) ligand. Through a salt metathesis reaction, the chloride ligand in 1 was replaced by potassium benzothiazole-2-thiolate (Kbt) and potassium benzoimidazole-2-thiolate (Kbi) to afford complexes (PhCH2NC)Au(κ1-S-bt)], 2a and (PhCH2NC)Au(κ1-S-bi)], 2b, respectively, which were characterized by NMR spectroscopy. The cytotoxic activities of 2a and 2b were evaluated against three human cancer cell lines, including A549 (lung), SKOV3 (ovary), and MCF-7 (breast). Our results indicated that 2a exhibited comparable cytotoxicity on investigated cell lines with cisplatin. It showed a good anti-proliferative activity with IC50 of 19.46, 11.76 and 13.27 μM against A549, SKOV3 and MCF-7 cell lines, respectively. The effects of these complexes on the proliferation of the non-tumorigenic epithelial breast cell line (MCF-10A) showed their good selectivity between the tumorigenic and non-tumorigenic cell lines. Molecular docking simulation studies were also conducted to determine the specific binding site and binding mode of the synthesized gold complexes to DNA and thioredoxinreductase (TrxR) as their proposed targets.
Sigma-Aldrich
Benzyl isocyanide, 98%