Merck
  • Molecular design of N-linked tetravalent glycosides bearing N-acetylglucosamine, N,N'-diacetylchitobiose and N-acetyllactosamine: Analysis of cross-linking activities with WGA and ECA lectins.
Molecular design of N-linked tetravalent glycosides bearing N-acetylglucosamine, N,N'-diacetylchitobiose and N-acetyllactosamine: Analysis of cross-linking activities with WGA and ECA lectins.
Bioorganic & medicinal chemistry (2010-01-09)
Ryuichi Masaka, Makoto Ogata, Yoshinori Misawa, Megumi Yano, Chika Hashimoto, Takeomi Murata, Hirokazu Kawagishi, Taichi Usui
ABSTRACT
Two types of nonspacer- and spacer-N-linked tetravalent glycosides bearing N-acetylglucosamine (GlcNAc), N,N'-diacetylchitobiose [(GlcNAc)(2)] and N-acetyllactosamine (LacNAc) were designed and prepared as glycomimetics. The interactions of wheat germ (Triticum vulgaris) agglutinin (WGA) and coral tree (Erythrina cristagalli) agglutinin (ECA) with a series of tetravalent glycosides and related compounds were studied using a hemagglutination inhibition assay, a precipitation assay, double-diffusion test, and an optical biosensor based on surface plasmon resonance (SPR). The tetravalent glycosides were found to be capable of binding and precipitating the lectins as tetravalent ligands. Strong interactions with WGA, due to a combination of multivalency effects and spacer effects, were observed for tetravalent glycosides bearing flexible tandem GlcNAc. The chelate effect leads to large rate enhancement for the tetravalent system with favorable orientation of ligands. Our simple strategy produced multivalent glycosides with strong cross-linking activity for lectin as a specific coagulant.
Sigma-Aldrich
N,N′-Diacetylchitobiose, ≥96% (HPLC)