Merck
  • Gamma radiation-induced preparation of chitosan-acrylic acid-1-vinyl-2-vinylpyrrolidone/multiwalled carbon nanotubes composite for removal of 152+154Eu, 60Co and 134Cs radionuclides.
Gamma radiation-induced preparation of chitosan-acrylic acid-1-vinyl-2-vinylpyrrolidone/multiwalled carbon nanotubes composite for removal of 152+154Eu, 60Co and 134Cs radionuclides.
International journal of biological macromolecules (2020-08-18)
Islam Mohamed Abdelmonem, Essam Metwally, Tharwat Essa Siyam, Farid Abou El-Nour, Abdel-Rahman Mahmoud Mousa
ABSTRACT
Removal behaviors of 152+154Eu, 60Co, and 134Cs radionuclides onto Chitosan-acrylic acid-1-vinyl-2-vinylpyrrolidone/oxidized multi-walled carbon nanotubes (CTS-AA-VP/o-MWCNTs) composite has been investigated by batch adsorption technique. CTS-AA-VP/o-MWCNTs composite has been synthesized by copolymerization of acrylic acid (AA) and 1-vinyl-2-vinylpyrrolidone (VP) onto the surface of chitosan/oxidized multi-walled carbon nanotubes (CTS/o-MWCNTs) using gamma radiation. SEM, TGA, and FTIR were applied to characterize the morphology, thermal stability, and structure of the composite. The composite shows high removal capacity of 321.77, 369.91, and 456.46 mg/g towards 152+154Eu, 60Co, and 134Cs radionuclides, respectively.
Sigma-Aldrich
Europium(III) oxide, 99.9% trace metals basis
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%