Merck
  • Aldehyde dehydrogenase 1A1 and gelsolin identified as novel invasion-modulating factors in conditioned medium of pancreatic cancer cells.
Aldehyde dehydrogenase 1A1 and gelsolin identified as novel invasion-modulating factors in conditioned medium of pancreatic cancer cells.
Journal of proteomics (2008-10-14)
Naomi Walsh, Paul Dowling, Norma O'Donovan, Michael Henry, Paula Meleady, Martin Clynes
ABSTRACT
Conditioned medium (CM) from clonal sub-populations of the pancreatic cancer cell line, MiaPaCa-2 with differing invasive abilities, were examined for their effect on in vitro invasion. Conditioned medium from Clone #3 (CM#3) strongly promoted invasion, while CM from Clone #8 (CM#8) inhibited invasion in vitro. 2D DIGE followed by MALDI-TOF MS analysis of CM#3 and CM#8 identified 41 proteins which were differentially regulated; 27 proteins were down-regulated and 14 proteins up-regulated in the invasion-promoting CM#3 when compared to CM#8. Western blotting analysis confirmed the down-regulated expression of gelsolin and the up-regulation of aldehyde dehydrogenase 1A1 in CM#3. Down-regulation of aldehyde dehydrogenase 1A1 in Clone #3 CM and gelsolin levels in Clone #8 CM by siRNA transfection revealed an important involvement of these proteins in promoting and inhibiting invasion in these pancreatic cancer cell lines.
Sigma-Aldrich
Monoclonal Anti-Gelsolin antibody produced in mouse, clone GS-2C4, ascites fluid