Merck
  • Plasma interferon-γ-inducible protein 10 (IP-10) levels correlate with disease severity and paradoxical reactions in extrapulmonary tuberculosis.
Plasma interferon-γ-inducible protein 10 (IP-10) levels correlate with disease severity and paradoxical reactions in extrapulmonary tuberculosis.
Infection (2020-11-04)
Isabelle Suárez, Samuel Rohr, Melanie Stecher, Clara Lehmann, Sandra Winter, Norma Jung, Vanessa Priesner, Melanie Berger, Christoph Wyen, Max Augustin, Jakob J Malin, Julia Fischer, Carola Horn, Florian Neuhann, Michael Püsken, Georg Plum, Gerd Fätkenheuer, Jan Rybniker
ABSTRACT
With 1.5 million deaths worldwide in 2018, tuberculosis (TB) remains a major global public health problem. While pulmonary TB (PTB) is the most common manifestation, the proportion of extrapulmonary TB (EPTB) is increasing in low-burden countries. EPTB is a heterogeneous disease entity posing diagnostic and management challenges due to the lack of reliable biomarkers. In this study, we prospectively evaluated clinical data and treatment response which were correlated with different biomarkers. The study was conducted at the University Hospital of Cologne. 20 patients with EPTB were enrolled. We analyzed plasma interferon-γ-inducible protein 10 (IP-10) levels in plasma by ELISA for up to 12 months of treatment. In addition, the QuantiFERON®-TB Gold Plus (QFT® Plus) test was performed during the course of treatment. Clinical data were assessed prospectively and correlated with QFT® Plus and IP-10 levels. Plasma IP-10 levels were found to be significantly increased (p < 0.001) in patients with extensive disease compared to patients with limited disease (cervical lymph node TB) or healthy controls. In patients with clinically confirmed paradoxical reaction (PR), a further increase of IP-10 was noted. IFN-γ measured by the QFT® Plus test did not decrease significantly during the course of treatment. Of note, in four EPTB patients (20%) without radiographic pulmonary involvement, sputum culture was positive for Mycobacterium tuberculosis. Our data demonstrate that IP-10 may be a valuable biomarker for estimation of disease severity in EPTB and monitoring of the disease course in extensive forms. However, IP-10 may be less suitable for diagnosis and monitoring of EPTB patients with limited disease. The QFT® Plus test does not appear to be a suitable marker for therapy monitoring. Sputum should be examined in EPTB patients even in case of normal diagnostic imaging of the chest.
Sigma-Aldrich
Human IP-10 / CXCL10 ELISA Kit, for serum, plasma, cell culture supernatant and urine