Merck
  • Selective inhibition of human immunodeficiency viruses by racemates and enantiomers of cis-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine.
Selective inhibition of human immunodeficiency viruses by racemates and enantiomers of cis-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine.
Antimicrobial agents and chemotherapy (1992-11-01)
R F Schinazi, A McMillan, D Cannon, R Mathis, R M Lloyd, A Peck, J P Sommadossi, M St Clair, J Wilson, P A Furman
ABSTRACT
2',3'-Dideoxy-5-fluoro-3'-thiacytidine (FTC) has been shown to be a potent and selective compound against human immunodeficiency virus type 1 in acutely infected primary human lymphocytes. FTC is also active against human immunodeficiency virus type 2, simian immunodeficiency virus, and feline immunodeficiency virus in various cell culture systems, including human monocytes. The antiviral activity can be prevented by 2'-deoxycytidine, but not by other natural nucleosides, suggesting that FTC must be phosphorylated to be active and 2'-deoxycytidine kinase is responsible for the phosphorylation. By using chiral columns or enzymatic techniques, the two enantiomers of FTC were separated. The (-)-beta-enantiomer of FTC was about 20-fold more potent than the (+)-beta-enantiomer against human immunodeficiency virus type 1 in peripheral blood mononuclear cells and was also effective in thymidine kinase-deficient CEM cells. Racemic FTC and its enantiomers were nontoxic to human lymphocytes and other cell lines at concentrations of up to 100 microM. Studies with human bone marrow cells indicated that racemic FTC and its (-)-enantiomer had a median inhibitory concentration of > 30 microM. The (+)-enantiomer was significantly more toxic than the (-)-enantiomer to myeloid progenitor cells. The susceptibilities to FTC of pretherapy isolates in comparison with those of posttherapy 3'-azido-3'-deoxythymidine-resistant viruses in human lymphocytes were not substantially different. Similar results were obtained with well-defined 2',3'-dideoxyinosine- and nevirapine-resistant viruses. (-)-FTC-5'-triphosphate competitively inhibited human immunodeficiency virus type 1 reverse transcriptase, with an inhibition constant of 2.9 microM, when a poly(I)n.oligo(dC)19-24 template primer was used. These results suggest that further development of the (-)-Beta-enantiomer of FTC is warranted as an antiviral agent for infections caused by human immunodeficiency viruses.
Sigma-Aldrich
Emtricitabine, ≥98% (HPLC)